Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tmem110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028839.4 | NP_083115.3 | 6, 7 | 752 | ||
GenBank mRNA | AK007740.1 | - | 6, 7 | 760 | ||
AK009341.1 | - | 6, 7 | 731 | |||
AK076341.1 | - | 6, 7 | 731 | |||
AK149088.1 | - | 6, 7 | 725 | |||
AK155257.1 | - | 6, 7 | 717 | |||
BC024583.1 | - | 4, 5 | 440 | 5065836 |