Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093752.1 | NP_001087221.1 | 4, 5 | 563 | ||
NM_001093753.1 | NP_001087222.1 | 3, 4 | 545 | |||
GenBank mRNA | AK011472.1 | - | 4, 5 | 501 | ||
AK029394.1 | - | 3, 4 | 508 | |||
AK081511.1 | - | 1, 2 | 370 | |||
AK146719.1 | - | 3, 4 | 517 | |||
AK215844.1 | - | 3, 4 | 357 | |||
AK218604.1 | - | 2, 3 | 135 | |||
BC002111.1 | - | 3, 4 | 516 | |||
BC021809.1 | - | 3, 4 | 500 | |||
BC037053.1 | - | 4, 5 | 486 | |||
BC038928.1 | - | 4, 5 | 522 | |||
BC055763.1 | - | 5, 6 | 451 | |||
BC069945.1 | - | 3, 4 | 495 | |||
BC138145.1 | - | 1, 2 | 298 | 8861164 | ||
BC145162.1 | - | 1, 2 | 298 | 9053713 | ||
BQ571667.1 | - | 1, 2 | 580 |