Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spaca4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027055.3 | NP_081331.2 | 1 | 516 | ||
GenBank mRNA | AK005760.1 | - | 1 | 508 | ||
BC048608.1 | - | 1 | 513 | 6742046 |