Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164827.1 | NP_001158299.1 | 1 | 31 | ||
NM_028054.3 | NP_082330.2 | 1 | 31 |
Gene Symbol: Dnajc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139139.2 | NP_631878.2 | 1 | 42 | ||
GenBank mRNA | BC013487.1 | - | 1 | 21 | 4211118 | |
BY794890.1 | - | 1 | 43 |