Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290381.1 | NP_001277310.1 | 5 | 870 | ||
NM_023524.2 | NP_076013.1 | 5 | 870 | |||
GenBank mRNA | AB035134.1 | - | 5 | 561 | ||
AF267988.1 | - | 5 | 825 | |||
AK010279.1 | - | 5 | 528 | |||
AK155925.1 | - | 5 | 854 | |||
AK177983.1 | - | 2 | 164 | |||
BC024356.1 | - | 5 | 769 | 5066740 | ||
CX768547.1 | - | 4 | 389 |