Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166503.1 | NP_001159975.1 | 10 | 1132 | ||
NM_027216.5 | NP_081492.4 | 10 | 1111 | |||
GenBank mRNA | AK007986.1 | - | 10 | 1051 | ||
AK036999.1 | - | 10 | 1058 | |||
AK049368.1 | - | 10 | 1076 | |||
AK165545.1 | - | 8 | 963 | |||
AK170673.1 | - | 10 | 1051 | |||
BC004643.1 | - | 6 | 641 | |||
BC019647.1 | - | 10 | 1055 | 3978277 | ||
BC031751.1 | - | 11 | 1169 | 4216487 |