Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700028J19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006462.1 | - | 3 | 303 | ||
BC049653.1 | - | 3 | 268 | |||
BC061239.1 | - | 2 | 183 | |||
BC087938.1 | - | 2 | 183 | |||
BC100477.1 | - | 3 | 284 |
Gene Symbol: Gm10109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131849.1 | - | 271 |