Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199955.1 | NP_001186884.1 | 1, 2 | 358 | ||
NM_027303.1 | NP_081579.1 | 1, 2 | 358 | |||
GenBank mRNA | AK006104.1 | - | 1, 2 | 294 | ||
AK006546.1 | - | 1, 2 | 300 | |||
BY706216.1 | - | 1, 2 | 290 |