Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083884.1 | NP_001077353.1 | 5 | 426 | ||
NM_027339.2 | NP_081615.2 | 4 | 361 | |||
GenBank mRNA | AK008940.1 | - | 5 | 415 | ||
AK136583.1 | - | 352 | ||||
BC026828.1 | - | 4 | 336 | 4218333 |