Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027357.2 | NP_081633.1 | 20 | 2454 | ||
GenBank mRNA | AK010596.1 | - | 4 | 400 | ||
AK087947.1 | - | 20 | 2405 | |||
AK153386.1 | - | 20 | 2455 | |||
AK159123.1 | - | 20 | 2454 | |||
AK159750.1 | - | 20 | 2433 | |||
AK180030.1 | - | 2 | 156 | |||
AK202172.1 | - | 2 | 156 | |||
AK219841.1 | - | 2 | 156 | |||
BC018172.1 | - | 6 | 503 | |||
BC022955.1 | - | 20 | 2344 | |||
BC025802.1 | - | 20 | 2358 | |||
BC029319.1 | - | 20 | 2391 | |||
BC031587.1 | - | 20 | 2385 | |||
BC032068.1 | - | 20 | 2385 | |||
BC039937.1 | - | 20 | 2431 | |||
BC089028.1 | - | 20 | 2401 | |||
BC138526.1 | - | 20 | 2417 | 8861546 | ||
BC138527.1 | - | 20 | 2417 | 8861547 |