Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly6g6e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027366.1 | NP_081642.1 | 1 | 239 | ||
GenBank mRNA | AJ315550.1 | - | 1 | 26 | ||
AJ315551.1 | - | 1 | 26 | |||
AK009303.1 | - | 1 | 96 | |||
BC132420.1 | - | 1 | 87 | 40130697 | ||
BC138777.1 | - | 1 | 87 | 8861799 | ||
BY238804.1 | - | 1 | 244 |