Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mier2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027422.2 | NP_081698.2 | 13, 14 | 1604 | ||
GenBank mRNA | AK012576.1 | - | 1, 2 | 453 | ||
AK154672.1 | - | 13, 14 | 1621 | |||
AK173128.1 | - | 12, 13 | 1759 | |||
BC005748.1 | - | 12 | ||||
BC067013.1 | - | 13, 14 | 1604 | 6810181 | ||
BC080757.1 | - | 6, 7 | 855 | |||
BC083321.1 | - | 4, 5 | 725 |