Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081242.2 | NP_001074711.2 | 12 | 1387 | ||
GenBank mRNA | AF467081.1 | - | 10 | 1113 | ||
AK165499.1 | - | 6 | 559 | |||
BC046480.1 | - | 4 | 359 | |||
BC079896.1 | - | 3 | 347 | 30544295 | ||
BC158057.1 | - | 12 | 1358 | 9007357 |