Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205236.1 | NP_001192165.1 | 3 | 848 | ||
NM_183288.3 | NP_899111.2 | 3 | 848 | |||
GenBank mRNA | AK017636.1 | - | 3 | 541 | ||
AK154926.1 | - | 3 | 849 | |||
BC138292.1 | - | 1 | 304 | 8861311 | ||
BC138293.1 | - | 1 | 304 | 8861312 |
Gene Symbol: Arhgap27os2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052773.1 | - | 2 | 918 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC048412.1 | - | 623 | Chr. - - [-] on Build |