Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133911.1 | NP_598672.1 | 19 | 3073 | ||
GenBank mRNA | AK028929.1 | - | 19 | 3012 | ||
AK131924.1 | - | 8 | 1215 | |||
AK195901.1 | - | 2 | 210 | |||
AK197812.1 | - | 2 | 210 | |||
AK210870.1 | - | 2 | 208 | |||
BC019649.1 | - | 8 | 1200 | |||
BC020317.2 | - | 14 | 2206 | |||
BC048928.1 | - | 3 | 343 | |||
BC052391.1 | - | 19 | 3073 | |||
BC058251.1 | - | 3 | 343 |
Set Membership: |
![]() |