Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027819.2 | NP_082095.2 | 4 | 669 | ||
GenBank mRNA | AK018485.1 | - | 4 | 671 | ||
BC055837.1 | - | 2 | 344 | 4225313 | ||
BC058747.1 | - | 4 | 669 | 6394797 |