Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023625.4 | NP_076114.2 | 4 | 634 | ||
GenBank mRNA | AF217003.1 | - | 4 | 615 | ||
AK004973.1 | - | 4 | 637 | |||
AK029813.1 | - | 4 | 634 | |||
AK030234.1 | - | 4 | 586 | |||
AK032930.1 | - | 4 | 630 | |||
AK042089.1 | - | 630 | ||||
AK043608.1 | - | 4 | 624 | |||
AK046937.1 | - | 4 | 624 | |||
AK150201.1 | - | 4 | 625 | |||
AK164220.1 | - | 4 | 615 | |||
AK170387.1 | - | 4 | 626 | |||
AK170631.1 | - | 4 | 614 | |||
AK194379.1 | - | 1 | 41 | |||
AK212536.1 | - | 1 | 41 | |||
BC026395.1 | - | 4 | 612 | |||
BC038605.1 | - | 4 | 615 | 4921245 | ||
BC048826.1 | - | 4 | 621 | 4196604 | ||
BC060095.1 | - | 4 | 612 |