Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Isyna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023627.1 | NP_076116.1 | 4, 5 | 482 | ||
GenBank mRNA | AF288525.1 | - | 4, 5 | 482 | ||
AK005029.2 | - | 4, 5 | 460 | |||
AK079323.1 | - | 4, 5 | 459 | |||
AK195643.1 | - | 1, 2 | 62 | |||
BC003458.1 | - | 4, 5 | 473 | 2648362 |