Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700008I05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027952.3 | NP_082228.2 | 6, 7 | 791 | ||
GenBank mRNA | AK005772.1 | - | 6, 7 | 792 | ||
BC141320.1 | - | 6, 7 | 772 | 9055932 | ||
BC141322.1 | - | 6, 7 | 772 | 9055934 |