Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079865.2 | NP_001073334.2 | 7, 8 | 1081 | ||
GenBank mRNA | AK040349.1 | - | 7, 8 | 1083 | ||
BC117742.1 | - | 7, 8 | 1081 | 40090393 | ||
BC117743.1 | - | 5, 6 | 688 | 40090395 |