Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tom1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028011.2 | NP_082287.1 | 14 | 1381 | ||
GenBank mRNA | AF395837.1 | - | 14 | 1345 | ||
AJ505007.1 | - | 14 | 1342 | |||
AK009826.1 | - | 12 | 1116 | |||
AK033712.1 | - | 14 | 1383 | |||
AK083630.1 | - | 1343 | ||||
BC004710.1 | - | 7 | 1738 | 3499918 |