Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ist1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028018.2 | NP_082294.1 | 6 | 618 | ||
GenBank mRNA | AK010262.1 | - | 6 | 588 | ||
AK050468.1 | - | 6 | 585 | |||
AK088192.1 | - | 6 | 573 | |||
AK122215.1 | - | 4 | 392 | |||
AK152830.1 | - | 6 | 585 | |||
AK161899.1 | - | 6 | 574 | |||
AK207302.1 | - | 4 | 312 | |||
AK218302.1 | - | 4 | 250 | |||
BC017605.1 | - | 6 | 584 | 4507347 | ||
BC039052.1 | - | 6 | 561 | 2631157 |