Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r14d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290796.1 | NP_001277725.1 | 1 | 199 | ||
NM_028104.4 | NP_082380.1 | 1 | 199 | |||
GenBank mRNA | AF408400.2 | - | 1 | 198 | ||
AK008348.1 | - | 1 | 157 | |||
AY179331.1 | - | 1 | 186 | |||
BC024547.1 | - | 1 | 198 | 4989982 |