Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162996.1 | NP_001156468.1 | 18 | 1769 | ||
NM_028153.1 | NP_082429.1 | 18 | 1826 | |||
GenBank mRNA | AK005562.1 | - | 18 | 1792 | ||
BC055476.1 | - | 18 | 1769 | 4218133 |