Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033165.3 | NP_001028337.2 | 2 | 419 | ||
GenBank mRNA | AK008359.1 | - | 3 | 173 | ||
AK008425.1 | - | 3 | 449 | |||
AK136359.1 | - | 2 | 113 | |||
BC119039.1 | - | 2 | 184 | 8733788 | ||
BC120693.1 | - | 2 | 184 | 40129616 |