Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tmx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028339.1 | NP_082615.1 | 8 | 794 | ||
GenBank mRNA | AK030429.1 | - | 8 | 812 | ||
AK050567.1 | - | 8 | 793 | |||
AK137059.1 | - | 8 | 791 | |||
AK150511.1 | - | 8 | 801 | |||
AK153027.1 | - | 8 | 797 | |||
AK170468.1 | - | 8 | 812 | |||
BC017603.1 | - | 8 | 794 | 4503129 | ||
BC021533.1 | - | 8 | 815 | 5354337 |