Gene Symbol: Ppp2r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028392.3 | NP_082668.1 | 8 | 1226 | ||
GenBank mRNA | AF512670.1 | - | 8 | 932 | ||
AF536771.1 | - | 8 | 995 | |||
AK013600.1 | - | 8 | 1228 | |||
AK018119.1 | - | 8 | 1189 | |||
AK039592.1 | - | 8 | 1069 | |||
AK079059.1 | - | 1676 | ||||
AK148956.1 | - | 8 | 1197 | |||
BC026686.1 | - | 8 | 1164 | 5065516 | ||
BC066007.1 | - | 8 | 1467 | |||
BC085147.1 | - | 8 | 1379 | |||
BC088979.1 | - | 8 | 1161 | 5690600 |