Gene Symbol: Ppp2r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028392.3 | NP_082668.1 | 9 | 1498 | ||
GenBank mRNA | AF512670.1 | - | 9 | 1204 | ||
AF536771.1 | - | 9 | 1267 | |||
AK013600.1 | - | 9 | 1500 | |||
AK018119.1 | - | 9 | 1461 | |||
AK039592.1 | - | 9 | 1341 | |||
AK148956.1 | - | 9 | 1469 | |||
BC026686.1 | - | 9 | 1436 | 5065516 | ||
BC066007.1 | - | 9 | 1739 | |||
BC085147.1 | - | 9 | 1651 | |||
BC088979.1 | - | 9 | 1433 | 5690600 |