Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Atat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142745.1 | NP_001136217.1 | 11 | 1021 | ||
NM_028476.4 | NP_082752.3 | 10 | 952 | |||
GenBank mRNA | AK014265.1 | - | 11 | 995 | ||
AK034725.1 | - | 10 | 930 | |||
AK042804.1 | - | 5 | 448 | |||
AK049682.1 | - | 10 | 902 | |||
AK133694.1 | - | 11 | 998 |