Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148940.3 | NP_683742.2 | 5 | 1055 | ||
GenBank mRNA | AB162857.1 | - | 5 | 1041 |