Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024261.2 | NP_077223.2 | 5 | 885 | ||
GenBank mRNA | BC099894.1 | - | 4 | 3641 | ||
BC138908.1 | - | 5 | 845 | 8861930 | ||
BC145732.1 | - | 5 | 845 | 40131035 |