Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cst6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028623.5 | NP_082899.1 | 1 | 445 | ||
GenBank mRNA | AK003744.1 | - | 1 | 257 | ||
AK078116.1 | - | 1 | 446 | |||
BC027680.1 | - | 1 | 198 | |||
BC061036.1 | - | 1 | 231 | 6770352 |