Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otud4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081164.1 | NP_001074633.1 | 2, 3 | 465 | ||
NM_001256033.1 | NP_001242962.1 | 2, 3 | 465 | |||
GenBank mRNA | AW494258.1 | - | 2, 3 | 465 | ||
BC138373.1 | - | 2, 3 | 437 | 8861393 | ||
BC145259.1 | - | 2, 3 | 437 | 9053812 |