Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pop7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028753.2 | NP_083029.1 | 2 | 570 | ||
GenBank mRNA | AK002782.1 | - | 2 | 572 | ||
AK158620.1 | - | 1 | 482 | |||
AK162268.1 | - | 2 | 575 | |||
BC010780.1 | - | 2 | 484 | 4216163 |