Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028810.2 | NP_083086.1 | 5 | 749 | ||
GenBank mRNA | AK011442.1 | - | 5 | 747 | ||
AK035195.1 | - | 5 | 734 | |||
AK048635.1 | - | 4 | 2284 | |||
BC009002.1 | - | 5 | 719 | 3500586 |