Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hopx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159900.1 | NP_001153372.1 | 2 | 192 | ||
NM_001159901.1 | NP_001153373.1 | 1 | 320 | |||
NM_175606.3 | NP_783199.1 | 2 | 352 | |||
GenBank mRNA | AF492703.1 | - | 2 | 352 | ||
AF492704.1 | - | 2 | 180 | |||
AF534182.1 | - | 2 | 397 | |||
AF536202.1 | - | 1 | 89 | |||
AI604115.1 | - | 1 | 306 | |||
AK003784.1 | - | 2 | 181 | |||
AK008297.1 | - | 2 | 159 | |||
AK008439.1 | - | 2 | 159 | |||
AK009007.1 | - | 2 | 109 | |||
BC024546.1 | - | 2 | 176 | 4989797 |
Gene Symbol: E130218H05 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087463.1 | - | 1 | 415 |