Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029031.3 | NP_083307.1 | 3 | 362 | ||
GenBank mRNA | AK015273.1 | - | 3 | 385 | ||
BC054721.1 | - | 3 | 362 | 5694588 | ||
BC056433.1 | - | 3 | 362 | 5694588 |