Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmbr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029098.3 | NP_083374.1 | 8 | 827 | ||
GenBank mRNA | AK003656.1 | - | 8 | 759 | ||
AK030589.1 | - | 8 | 793 | |||
AK049110.1 | - | 8 | 828 | |||
AK143891.1 | - | 8 | 788 | |||
AK154968.1 | - | 8 | 961 | |||
AK170328.1 | - | 8 | 788 | |||
AK173121.1 | - | 8 | 1208 | |||
AY052398.1 | - | 8 | 670 | |||
BC023397.1 | - | 8 | 764 | 5043877 |