Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam228a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029107.2 | NP_083383.1 | 4 | 509 | ||
GenBank mRNA | AK015158.1 | - | 4 | 381 | ||
AK029492.1 | - | 4 | 510 | |||
AK133272.1 | - | 4 | 546 | |||
BC138846.1 | - | 4 | 444 | 8861868 | ||
BC138853.1 | - | 4 | 352 | 8861875 |