Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930523C07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162896.1 | NP_001156368.1 | 3 | 662 | ||
GenBank mRNA | AK028922.1 | - | 3 | 464 | ||
AK034159.1 | - | 3 | 358 | |||
AK035814.1 | - | 3 | 657 |
Gene Symbol: 4930562F07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016191.1 | - | 3 | 348 | ||
BU938572.1 | - | 278 |