Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930523C07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162896.1 | NP_001156368.1 | 3 | 3255 | ||
GenBank mRNA | AK028660.1 | - | 1294 | |||
AK054156.1 | - | 852 | ||||
BC030429.1 | - | 1 | 73 |
Gene Symbol: 4930562F07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016191.1 | - | 1 | 109 |