Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syce3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162880.1 | NP_001156352.1 | 3 | 227 | ||
NM_001162882.1 | NP_001156354.1 | 3 | 195 | |||
GenBank mRNA | AK005702.1 | - | 3 | 149 | ||
AK029781.1 | - | 3 | 196 | |||
BC048615.1 | - | 3 | 227 | |||
EU079025.1 | - | 3 | 188 | |||
HQ130280.1 | - | 2 | 122 |