Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145451.3 | NP_663426.2 | 5 | 577 | ||
GenBank mRNA | AK006161.1 | - | 3 | 193 | ||
CB955421.1 | - | 5 | 601 | |||
CD363995.1 | - | 4 | 425 |