Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080808.1 | NP_001074277.1 | 9 | 1709 | ||
GenBank mRNA | AK170295.1 | - | 3 | 318 | ||
BC024804.1 | - | 9 | 1702 | |||
BC049161.1 | - | 3 | 447 | |||
BC064124.1 | - | 1 | 365 | |||
BC126948.1 | - | 9 | 1709 | 40090576 | ||
GU973876.1 | - | 9 | 1709 |