Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177077.2 | NP_796051.2 | 16, 17 | 1778 | ||
GenBank mRNA | AK144547.1 | - | 16, 17 | 1779 | ||
AK165019.1 | - | 15, 16 | 2398 | |||
AK220360.1 | - | 1, 2 | 52 | |||
BC059244.1 | - | 7, 8 | 689 | |||
BC144781.1 | - | 16, 17 | 1721 | 9053329 | ||
BC145693.1 | - | 16, 17 | 1721 | 40130996 |