Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198006.4 | NP_932123.3 | 2, 3 | 321 | ||
GenBank mRNA | AK050432.1 | - | 2, 3 | 227 | ||
AK166651.1 | - | 2, 3 | 251 | |||
AK169639.1 | - | 2, 3 | 263 | |||
AK177444.1 | - | 2, 3 | 142 | |||
AK218336.1 | - | 2, 3 | 142 | |||
BB860009.1 | - | 2, 3 | 272 | |||
BB863739.1 | - | 2, 3 | 264 | |||
BC002137.1 | - | 2, 3 | 228 | 3484305 |