Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029711.1 | NP_083987.1 | 7 | 935 | ||
GenBank mRNA | AK008777.1 | - | 3 | 381 | ||
AK083263.1 | - | 7 | 745 | |||
AK151513.1 | - | 7 | 909 | |||
AK152356.1 | - | 8 | 760 | |||
AK170061.1 | - | 8 | 734 | |||
AK211266.1 | - | 1 | 36 | |||
BC099504.1 | - | 6 | 587 | |||
BC115750.1 | - | 7 | 733 | 40051600 |