Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081379.2 | NP_001074848.2 | 5 | 837 | ||
GenBank mRNA | AK164512.1 | - | 2 | 3278 | ||
BC005576.1 | - | 3 | 434 | |||
BC158043.1 | - | 4 | 502 | 9088159 | ||
BC172141.1 | - | 5 | 599 | 9054811 |