Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029921.1 | NP_084197.1 | 1, 2 | 201 | ||
GenBank mRNA | AK021320.1 | - | 1, 2 | 202 | ||
BC104374.1 | - | 1, 2 | 193 | 40049947 | ||
BC104375.1 | - | 1, 2 | 193 | 40049948 | ||
BC119477.1 | - | 1, 2 | 193 | 40049949 |