Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030083.2 | NP_084359.1 | 2 | 312 | ||
GenBank mRNA | AK018782.1 | - | 2 | 313 | ||
BC038659.1 | - | 2 | 266 | |||
BC048499.1 | - | 2 | 318 | 6583055 | ||
BC062267.1 | - | 2 | 282 | 1532489 |